Merck
Inter- and intra-laboratory testing of the Daphnia magna IQ toxicity test.
Bulletin of environmental contamination and toxicology (1996-10-01)
K R Hayes, W S Douglas, J Fischer
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Methylumbelliferyl α-D-Galactopyranosid, ≥98% (TLC)
Sigma-Aldrich
4-Methylumbelliferyl β-D-Galactopyranosid, ≥99% (TLC)