Merck
Ontogeny of sulphated glycoconjugate-producing cells in the rat fundic gland.
The Histochemical journal (1996-01-01)
D H Yang, H Kasamo, M Miyauchi, S Tsuyama, F Murata
ZUSAMMENFASSUNG
The ontogeny of sulphated glycoconjugate-producing cells in the rat fundic gland has been studied using high iron diamine (HID), Alcian Blue (AB) at pH 1.0, high iron diamine in combination with Alcian Blue at pH 2.5 (HID-AB), cationic colloidal gold (CCG) at pH 1.0 under light microscopy and CCG (1.0), HID-thiocarbohydrazide (TCH)-silver proteinate (SP)-physical development (PD) under electron microscopy. From day 19.5 of gestation, sulphated glycoconjugate-producing cells were discernible under both light and electron microscopy. The development of such cells can be classified into four stages: (1) a prenatal period from day 19.5 of gestation extending to 0.5 days after birth; (2) 1 day to 2 weeks after birth; (3) 2 to 4 weeks after birth; and (4) the final period from 4 to 8 weeks after birth. Glycoconjugate-producing cells reached maturity by 4 weeks after birth. Our results indicated that glycoconjugate-producing cells were cells along the wall of foveolar lumen, but not those covering the gastric mucosa surface. Our results also suggested that the trans to transmost Golgi apparatus lamellae were the sites of sulphation in the developing rat stomach.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thiocarbohydrazid, 98%