- Physicochemical data for some selected Fusarium toxins.
Physicochemical data for some selected Fusarium toxins.
Journal of AOAC International (1996-11-01)
E W Sydenham, P G Thiel, R Vleggaar
PMID8946715
ZUSAMMENFASSUNG
Fusarium toxins are a major group of secondary metabolites, produced by several species, that may contaminate food cereals and animal feeds. We describe results of a study in which a number of physicochemical constants for 12 Important Fusarium mycotoxins (zearalenone, diacetoxyscirpenol, T-2 toxin, neosolaniol monoacetate, deoxynivalenol, nivalenol, fumonisin B1, fumonisin B2, moniliformin, fusarenon-X, HT-2 toxin, and beta-zearalenol) were determined. Nuclear magnetic resonance, mass spectrometric, UV spectral, molar absorption coefficients, fluorescence spectra, melting points, and specific rotation data are presented.