Merck
Contact allergy to polyvinylpyrrolidone/hexadecene copolymer previously reported.
American journal of contact dermatitis : official journal of the American Contact Dermatitis Society (1996-12-01)
A C De Groot
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Hexadecen, ≥98.5%
Supelco
1-Hexadecen, analytical standard