Merck
Ultrastructural localization of sympathetic axons in experimental rat sciatic nerve neuromas.
Journal of neurocytology (1996-10-01)
J R Small, J W Scadding, D N Landon
ZUSAMMENFASSUNG
The ultrastructural localization of sympathetic axons was investigated in normal rat sciatic nerves and experimental sciatic nerve neuromas. The best ultrastructural localization of noradrenaline in the dense-cored vesicles of sympathetic axons was accomplished following pretreatment of rats with nialamide and 5-hydroxy dopamine, followed by fixation according to the modified chromaffin technique of Tranzer and Richards (1976). After such preparation, sympathetic axons containing 5-hydroxy dopamine-labelled dense-cored vesicles could be identified in normal sciatic nerve. Large accumulations of labelled dense-cored vesicles were also found in acute neuromas, up to 1 week after nerve section. Much smaller numbers of dense-cored vesicles could be identified in chronic neuromas from 2 to 3 weeks following nerve section. Sympathetic axons could also be identified following electron probe X-ray microanalysis of the tissue sections, using chromium detection as the marker for the noradrenaline-containing dense-cored vesicles. Unusual configurations of Schwann cell subunits, which enclosed myelinated fibres and sympathetic axon sprouts within the same basal lamina, were identified in the acute neuromas, 3-7 days after nerve section. Such configurations may be of relevance to the pathophysiological interaction which develops between sympathetic efferent and sensory fibres in peripheral nerve neuromas.