Merck
  • Simultaneous determination of acrolein, malonaldehyde and 4-hydroxy-2-nonenal produced from lipids oxidized with Fenton's reagent.
Simultaneous determination of acrolein, malonaldehyde and 4-hydroxy-2-nonenal produced from lipids oxidized with Fenton's reagent.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (1996-10-01)
T Miyake, T Shibamoto
ZUSAMMENFASSUNG
Ethyl linoleate, ethyl linolenate, ethyl arachidonate and cod liver oil were oxidized with Fenton's reagent. Acrolein, malonaldehyde and 4-hydroxynonenal formed were derivatized to N-methylpyrazoline, N-methylpyrazole and 5-(1'-hydroxyhexyl)-1-methyl-2-pyrazoline with N-methylhydrazine, respectively. The derivatives were simultaneously analysed by gas chromatograph equipped with a fused silica capillary column and a nitrogen-phosphorus detector. The maximum amounts of acrolein (9.7 +/- 2.11 nmol/ml) and malonaldehyde (61.18 +/- 6.51 nmol/ml) were formed from cod liver oil. The highest amount of 4-hydroxynonenal (6.83 +/- 0.53 nmol/ml) was produced from ethyl arathidonate.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyllinoleat, ≥99%
Sigma-Aldrich
Ethyl-Linolenat, ≥98%
Sigma-Aldrich
Ethyllinoleat, technical, ≥65% (GC)
Sigma-Aldrich
Ethyl arachidonate, ≥98.5% (GC), liquid