Merck
Green hair discoloration due to selenium sulfide.
International journal of dermatology (1997-03-01)
E A Fitzgerald, S M Purcell, H M Goldman
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Selensulfid