Merck
  • p-Hydroxyphenylacetaldehyde, the major product of L-tyrosine oxidation by the myeloperoxidase-H2O2-chloride system of phagocytes, covalently modifies epsilon-amino groups of protein lysine residues.
p-Hydroxyphenylacetaldehyde, the major product of L-tyrosine oxidation by the myeloperoxidase-H2O2-chloride system of phagocytes, covalently modifies epsilon-amino groups of protein lysine residues.
The Journal of biological chemistry (1997-07-04)
S L Hazen, J P Gaut, F F Hsu, J R Crowley, A d'Avignon, J W Heinecke
ZUSAMMENFASSUNG
Activated human phagocytes employ the myeloperoxidase-H2O2-Cl- system to convert L-tyrosine to p-hydroxyphenylacetaldehyde (pHA). We have explored the possibility that pHA covalently reacts with proteins to form Schiff base adducts, which may play a role in modifying targets at sites of inflammation. Because Schiff bases are labile to acid hydrolysis, prior to analysis the adducts were rendered stable by reduction with NaCNBH3. Purified pHA reacted with Nalpha-acetyllysine, an analog of protein lysine residues. The reduced reaction product was identified as Nalpha-acetyl-Nepsilon-(2-(p-hydroxyphenyl)ethyl)lysine by 1H NMR spectroscopy and mass spectrometry. The compound Nepsilon-(2-(p-hydroxyphenyl)ethyl)lysine (pHA-lysine) was likewise identified in acid hydrolysates of bovine serum albumin (BSA) that were first exposed to myeloperoxidase, H2O2, L-tyrosine, and Cl- and then reduced with NaCNBH3. Other halides (F-, Br-, I-) and the pseudohalide SCN- could not replace Cl- as a substrate in the myeloperoxidase-H2O2-L-tyrosine system. In the absence of the enzymatic system, pHA-lysine was detected in reduced reaction mixtures of BSA, L-tyrosine, and reagent HOCl. In contrast, pHA-lysine was undetectable when BSA was incubated with L-tyrosine and HOBr, peroxynitrite, hydroxyl radical, or a variety of other peroxidases, indicating that the aldehyde-protein adduct was selectively produced by HOCl. Human neutrophils activated in the presence of tyrosine also modified BSA lysine residues. pHA-lysine formation required L-tyrosine and cell activation; it was inhibited by peroxidase inhibitors and catalase, implicating myeloperoxidase and H2O2 in the reaction pathway. pHA-lysine was detected in inflamed human tissues that were reduced, hydrolyzed, and then analyzed by mass spectrometry, indicating that the reaction of pHA with proteins may be of physiological importance. These observations raise the possibility that the identification of pHA-lysine in tissues will pinpoint targets where phagocytes inflict oxidative damage in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nε-Acetyl-L-lysin