- Novel substrates of yeast alcohol dehydrogenase-3. 4-dimethylamino-cinnamaldehyde and chloroacetaldehyde.
Novel substrates of yeast alcohol dehydrogenase-3. 4-dimethylamino-cinnamaldehyde and chloroacetaldehyde.
Biochemistry and molecular biology international (1997-11-14)
V Leskovac, S Trivić, J Zeremski, B Stancić, B M Anderson
PMID9350344
ZUSAMMENFASSUNG
4-Dimethylamino-trans-cinnamaldehyde and chloroacetaldehyde are novel substrates of yeast alcohol dehydrogenase (EC 1.1.1.1). In the present work, we have reported the steady-state kinetic constants for both substrates, and their chemical reactions with the enzyme protein itself. Both substrates are potentially useful for biotechnology, chemoenzyme syntheses and analytical biochemistry.