Merck
  • Phenytoin hydroxylation in a healthy Caucasian population: bimodal distribution of hydroxyphenytoin urinary excretion.
Phenytoin hydroxylation in a healthy Caucasian population: bimodal distribution of hydroxyphenytoin urinary excretion.
Pharmacology & toxicology (1998-01-28)
Y Horsmans, V Van den Berge, A Bouckaert, J P Desager
ZUSAMMENFASSUNG
The phenytoin hydroxylation index was studied in 122 unrelated Caucasian volunteers. Following a 100 mg oral dose of phenytoin, phenytoin and hydroxyphenytoin were measured in urine from 0-32 hr after administration. As phenytoin was not found in all urine collections, a phenytoin hydroxylation index was expressed as follows: Phenytoin hydroxylation index = amount of phenytoin administered/0-32 hr urinary output of hydroxyphenytoin. Phenytoin hydroxylation index values appear to be bimodally distributed, 92% of the population showing a mean (+/- S.E.M.) value of 0.639 +/- 0.099 and 8% a mean (+/- S.E.M.) value of 1.001 +/- 0.180 (log10 values). These results are in favour of the existence of a phenytoin genetic polymorphism. Since misuse of urinary metabolite excretion data in drug metabolism studies is a well-known phenomenon, our data emphasize the need for future population studies on phenytoin pharmacokinetics as well as on CYP2C9 genotyping before concluding about existence of a phenytoin genetic polymorphism.