Merck
  • 3-Amino-5-hydroxybenzoic acid synthase, the terminal enzyme in the formation of the precursor of mC7N units in rifamycin and related antibiotics.
3-Amino-5-hydroxybenzoic acid synthase, the terminal enzyme in the formation of the precursor of mC7N units in rifamycin and related antibiotics.
The Journal of biological chemistry (1998-04-16)
C G Kim, T W Yu, C B Fryhle, S Handa, H G Floss
ZUSAMMENFASSUNG
The biosynthesis of ansamycin antibiotics, like rifamycin B, involves formation of 3-amino-5-hydroxybenzoic acid (AHBA) by a novel variant of the shikimate pathway. AHBA then serves as the starter unit for the assembly of a polyketide which eventually links back to the amino group of AHBA to form the macrolactam ring. The terminal enzyme of AHBA formation, which catalyzes the aromatization of 5-deoxy-5-amino-3-dehydroshikimic acid, has been purified to homogeneity from Amycolatopsis mediterranei, the encoding gene has been cloned, sequenced, and overexpressed in Escherichia coli. The recombinant enzyme, a (His)6 fusion protein, as well as the native one, are dimers containing one molecule of pyridoxal phosphate per subunit. Mechanistic studies showed that the enzyme-bound pyridoxal phosphate forms a Schiff's base with the amino group of 5-deoxy-5-amino-3-dehydroshikimic acid and catalyzes both an alpha, beta-dehydration and a stereospecific 1,4-enolization of the substrate. Inactivation of the gene encoding AHBA synthase in the A. mediterranei genome results in loss of rifamycin formation; production of the antibiotic is restored when the mutant is supplemented with AHBA.