Merck
The skin sensitization potential of four alkylalkanolamines.
Veterinary and human toxicology (1998-04-29)
H W Leung, D L Blaszcak
ZUSAMMENFASSUNG
The skin sensitization potential of 4 alkylalkanolamines (N-methylethanolamine, N,N-dimethylethanolamine, N-methyldiethanolamine and N,N-diethylethanolamine), was evaluated in a guinea pig maximation procedure by the method of Magnusson and Kligman. While all 4 alkylalkanolamines tested were irritating to the guinea pig skin, only N-methylethanolamine showed potential to induce allergic contact dermatitis. None of the remaining 3 alkylalkanolamines exhibited clear skin responses suggestive of sensitization.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Methyldiethanolamin, ≥99%
Sigma-Aldrich
2-(Methylamino)ethanol, ≥98%