Merck
An assessment of the genetic toxicology of antimony trioxide.
Mutation research (1998-08-26)
B M Elliott, J M Mackay, P Clay, J Ashby
ZUSAMMENFASSUNG
Antimony trioxide (Sb2O3, CAS 1309-64-4) has been examined in a range of in vitro and in vivo genotoxicity assays. Negative results were obtained with the Salmonella/microsome assay and the L5178Y mutation assay, but a positive response was observed in the in vitro cytogenetic assay using isolated human peripheral lymphocytes. However, in vivo, antimony trioxide was non-clastogenic in the mouse bone marrow micronucleus assay, following oral gavage administration for 1, 7, 14 or 21 days at dose levels of up to 5000 mg/kg (single dose) or 1000 mg/kg (repeat dose). A negative result was also obtained in the in vivo rat liver DNA repair (unscheduled DNA synthesis) assay following a single oral gavage administration of doses up to 5000 mg/kg. These data show no genotoxicity for antimony trioxide in vivo and do not confirm a previous report of clastogenicity in the mouse on repeated dosing. It is concluded that antimony trioxide is not genotoxic in vivo and does not present a genotoxic hazard to humans.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Antimon(III)-Oxid, powder, 5 μm, ReagentPlus®, 99%
Sigma-Aldrich
Antimon(III)-Oxid, 99.999% trace metals basis
Sigma-Aldrich
Antimon(III)-Oxid, 99.99% trace metals basis
Sigma-Aldrich
Antimon(III)-Oxid, nanopowder, <250 nm particle size (TEM), ≥99.9% trace metals basis