Merck
Dextran-70 inhibits equine platelet aggregation induced by PAF but not by other agonists.
Equine veterinary journal (1998-10-03)
M F Heath, R J Evans, L J Hayes
ZUSAMMENFASSUNG
Dextrans of mean molecular weight 70 kDa (dextran-70) have had clinical use as anti-thrombotics in man. A major part of the anti-thrombotic action is mediated via inhibition of platelet function. Greatorex (1975, 1977) treated thromboembolic colic in horses with infusions of dextran-70 and reported a 90% recovery rate, but this treatment is nonetheless rarely used. We have used an in vitro method to examine the effect of dextran-70 on equine platelet suspensions, in the hope that understanding the mechanism of action of dextran-70 might lead to the development of alternative therapeutic agents. The effects of dextran-70 on equine platelets occurred immediately in vitro with an initial activation and shape change. Subsequent assessment of aggregation revealed a dose-dependent specific inhibition of platelet-activating factor (PAF)-induced aggregation, significant in rate of aggregation at dextran-70 concentrations >40 g/l (P<0.05) and in extent of aggregation at dextran-70 concentrations >50 g/l (P<0.05). Pre-incubation with 60 g/l dextran-70 significantly inhibited the rate and extent of aggregation in response to PAF (1 nmol/l) (P<0.001 and P = 0.003, respectively) but this was not dependent on the duration of pre-incubation (from 0 to 150 min). No effects were seen when the agonist was adenosine 5'-diphosphate (200 nmol/l), collagen (10 mg/l), 5-hydroxytryptamine (100 micromol/l) or U44069 (600 nmol/l) (all P>0.1). Analysis of PAF concentration-aggregation curves after pre-incubation with 60 g/l dextran-70 indicated significant noncompetitive inhibition by dextran-70 (P<0.001 for rate and extent of aggregation). The ability of dextran-70 to inhibit responses of equine platelets to PAF is probably an important component of its beneficial effect as an anti-thrombotic in colic cases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
9,11-Dideoxy-9α,11α-epoxymethanoprostaglandin F