Merck
  • 20beta-hydroxysteroid dehydrogenase catalyzes ketone-reduction of acetohexamide, an oral antidiabetic drug, in liver microsomes of adult male rats.
20beta-hydroxysteroid dehydrogenase catalyzes ketone-reduction of acetohexamide, an oral antidiabetic drug, in liver microsomes of adult male rats.
The Journal of pharmacology and experimental therapeutics (1998-11-10)
H Takada, M Otagiri, Y Imamura
ZUSAMMENFASSUNG
We examined the catalytic properties and physiological function of an enzyme responsible for the ketone-reduction of acetohexamide, an oral antidiabetic drug, in liver microsomes of adult male rats. Progesterone, 17alpha-hydroxyprogesterone, cortisone and cortisol, which have a ketone group at 20-position of C21-steroids, were potent inhibitors for ketone-reduction of acetohexamide in liver microsomes of adult male rats. Progesterone was also found to inhibit competitively the ketone-reduction of acetohexamide, suggesting that the ketone-reduction of acetohexamide and progesterone is catalyzed by the same enzyme. When progesterone was used as a substrate, 20beta-hydroxysteroid dehydrogenase present in liver microsomes of adult rats, such as acetohexamide reductase, exhibited a male-specific and androgen-dependent activity. Furthermore, a significant correlation was observed between the activities of 20beta-hydroxysteroid dehydrogenase and acetohexamide reductase in liver microsomes of individual male rats at various ages. Based on all results, we conclude that 20beta-hydroxysteroid dehydrogenase catalyzes the ketone-reduction of acetohexamide in liver microsomes of adult male rats.