Merck
  • The alpha9 nicotinic acetylcholine receptor shares pharmacological properties with type A gamma-aminobutyric acid, glycine, and type 3 serotonin receptors.
The alpha9 nicotinic acetylcholine receptor shares pharmacological properties with type A gamma-aminobutyric acid, glycine, and type 3 serotonin receptors.
Molecular pharmacology (1999-02-03)
C V Rothlin, E Katz, M Verbitsky, A B Elgoyhen
ZUSAMMENFASSUNG
In the present study, we provide evidence that the alpha9 nicotinic acetylcholine receptor (nAChR) shares pharmacological properties with members of the Cys-loop family of receptors. Thus, the type A gamma-aminobutyric acid receptor antagonist bicuculline, the glycinergic antagonist strychnine, and the type 3 serotonin receptor antagonist ICS-205,930 block ACh-evoked currents in alpha9-injected Xenopus laevis oocytes with the following rank order of potency: strychnine > ICS-205,930 > bicuculline. Block by antagonists was reflected in an increase in the acetylcholine (ACh) EC50 value, with no changes in agonist maximal response or Hill coefficient, which suggests a competitive type of block. Moreover, whereas neither gamma-aminobutyric acid nor glycine modified ACh-evoked currents, serotonin blocked responses to ACh in a concentration-dependent manner. The present results suggest that the alpha9 nAChR must conserve in its primary structure some residues responsible for ligand binding common to other Cys-loop receptors. In addition, it adds further evidence that the alpha9 nAChR and the cholinergic receptor present at the base of cochlear outer hair cells have similar pharmacological properties.