- Spectrophotometric studies and application of imipramine-eriochrome cyanine R system for determination of imipramine in pharmaceuticals.
Spectrophotometric studies and application of imipramine-eriochrome cyanine R system for determination of imipramine in pharmaceuticals.
Journal of pharmaceutical and biomedical analysis (2000-08-10)
B Starczewska
PMID10933530
ZUSAMMENFASSUNG
Eriochrome cyanine R (ECR) has been tested as reagent for the determination of imipramine. It reacts in neutral medium with imipramine forming reddish compound, which can be quantitatively extracted into n-butanol. This property has been successfully used for the extractive-spectrophotometric determination of imipramine. Beer's law is obeyed in concentration range of 10-80 microg ml(-1) of imipramine. The method was applied to the determination of imipramine in its pharmaceutical.