Merck
  • Ion solvation and association in LiClO4/sulfolane solution: a vibrational spectroscopic and molecular orbital study.
Ion solvation and association in LiClO4/sulfolane solution: a vibrational spectroscopic and molecular orbital study.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2001-07-27)
X Xuan, J Wang, J Lu, N Pei, Y Mo
ZUSAMMENFASSUNG
Solvation interaction and ion association in solutions of lithium perchlorate/sulfolane have been studied by using infrared and Raman spectra as a function of concentration of lithium perchlorate. The band changes of antisymmetric OSO stretch, antisymmetric CSC stretch, -SO2 wag and twist suggest that there is an interaction between Li+ and sulfolane molecules, and the site of solvation is the oxygen atom of -SO2 group. The molecular orbital calculation supports this suggestion. On the other hand, the apparent solvation number was calculated, and the band fitting for the ClO4- band reveals the presence of contact ion pair, solvent separated ion pair and free ClO4- anion in the concentrated solutions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sulfolan, 99%
Supelco
Sulfolan, analytical standard