Merck
Effect of the herbicide 4-CPA on human erythrocyte antioxidant enzymes in vitro.
Redox report : communications in free radical research (2001-08-29)
Y Alicigüzel, S Ozdem, A Y Demir, F Unal, D Kumbul, S S Ozdem, G Perry, M A Smith
ZUSAMMENFASSUNG
To investigate the possible role of oxygen free radicals and oxidant stress in the toxic effects of phenoxyherbicides, we studied the in vitro effect of 4-chlorophenoxyacetic acid (4-CPA) on various human erythrocyte antioxidant enzymes, namely glucose-6-phosphate dehydrogenase, catalase, selenium-dependent glutathione peroxidase, glutathione reductase and Cu/Zn-superoxide dismutase. 4-CPA added in a dose of 1 ppm to human erythrocytes for 1 h caused a significant reduction in glucose-6-phosphate dehydrogenase (P <0.001) and catalase (P <0.001) activities, but did not significantly affect the activities of other enzymes. Such selective inactivation of specific erythrocyte antioxidant enzymes may play a role in the toxic effects of phenoxyherbicides.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Chlorphenoxyessigsäure, BioReagent, suitable for plant cell culture, crystalline
Sigma-Aldrich
4-Chlorphenoxyessigsäure, ≥98.0% (T)