Merck
[Studies on chemical constituents of Lysimachia pentapetala].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2003-02-11)
Z Zhang, Q Zhan
ZUSAMMENFASSUNG
From the ethanol extract of Lysimachia pentapetala, six compounds have been isolated. These compounds were identified as triacontanol, beta-sitosterol, glycerol monopalmitate, succinic acid, flavonol glycosides and hitrite upon the date of mp, IR, MS and 1HNMR tests.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Triacontanol, ≥98% (capillary GC)