Merck
  • Cell proliferation in the germinal layer of the lateral brain ventricles in normal adult mice and under some experimental conditions.
Cell proliferation in the germinal layer of the lateral brain ventricles in normal adult mice and under some experimental conditions.
Folia biologica (1992-01-01)
Z Srebro, I Płonkowa, E Pierścińska
ZUSAMMENFASSUNG
Histological and autoradiographic studies revealed mitotic activity in the subependymal germinal layer in the brains of adult mice. The number of mitoses observed was higher in animals subjected to administration of haematoporphyrin and selenourea.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Selenharnstoff, 98%