Merck
Vascular endothelial growth factor increases the intracellular magnesium.
Biochemical and biophysical research communications (2006-07-11)
Bing-Zhe Hong, Hyung-Sub Kang, June-No So, Han-Na Kim, Sun-Ah Park, Shang-Jin Kim, Kyung-Ryoul Kim, Yong-Geun Kwak
ZUSAMMENFASSUNG
Vascular endothelial growth factor (VEGF) is one of the key players in the process of angiogenesis. However, its underlying mechanism remains unclear. Mg2+ is the most abundant intracellular divalent cation in the body and plays critical roles in many cell functions. We investigated the effect of VEGF on intracellular Mg2+ in human umbilical vein endothelial cells (HUVECs). VEGF-A165 increased the intracellular Mg2+ concentration ([Mg2+]i) in a dose-dependent manner, with or without extracellular Mg2+, and the increase of [Mg2+]i was blocked by pretreatment with SU1498, tyrosine kinase inhibitors (tyrphostin A-23 and genistein), phosphatidylinositol 3-kinase (PI3K) inhibitors (wortmannin and LY294002) or phospholipase Cgamma (PLCgamma) inhibitor (U73122). In contrast, mitogen-activated protein kinase inhibitors (SB202190 and PD98059) had no effect on the VEGF-induced [Mg2+]i increase. These results suggest that VEGF-A165 increases the [Mg2+]i from the intracellular Mg2+ stores through the tyrosine kinase/PI3K/PLCgamma-dependent signaling pathways.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tyrphostin 23, ≥98%