Merck
  • Degradation of fenamiphos in soils collected from different geographical regions: the influence of soil properties and climatic conditions.
Degradation of fenamiphos in soils collected from different geographical regions: the influence of soil properties and climatic conditions.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2008-04-26)
Tanya Cáceres, Mallavarapu Megharaj, Ravi Naidu
ZUSAMMENFASSUNG
The persistence of fenamiphos (nematicide) in five soils collected from different geographical regions such as Australia, Ecuador and India under three temperature regimes (18, 25 and 37 degrees C) simulating typical environmental conditions was studied. The effect of soil properties (soil pH, temperature and microbial biomass) on the degradation of fenamiphos was determined. The rate of degradation increased with increase in temperature. Fenamiphos degradation was higher at 37 degrees C than at 25 and 18 degrees C (except under alkaline pH). The degradation pathway differed in different soils. Fenamiphos sulfoxide (FSO) was identified as the major degradation product in all the soils. Fenamiphos sulfone (FSO2), and the corresponding phenols: fenamiphos phenol (FP), fenamiphos sulfoxide phenol (FSOP) and fenamiphos sulfone phenol (FSO2P) were also detected. The degradation of fenamiphos was faster in the alkaline soils, followed by neutral and acidic soils. Under sterile conditions, the dissipation of the pesticide was slower than in the non-sterile soils suggesting microbial role in the pesticide degradation. The generation of new knowledge on fenamiphos degradation patterns under different environmental conditions is important to achieve better pesticide risk management.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Fenamiphos, PESTANAL®, analytical standard