- Simulated rainfall removal of tricyclazole sprayed on rice foliage.
Simulated rainfall removal of tricyclazole sprayed on rice foliage.
Bulletin of environmental contamination and toxicology (2008-05-20)
Thai Khanh Phong, Dang Thi Tuyet Nhung, Kenichi Yamazaki, Kazuhiro Takagi, Hirozumi Watanabe
PMID18488130
ZUSAMMENFASSUNG
Two rainfall simulations of 30 mm h(-1), with 48-h interval between two simulations, were performed on rice lysimeters at 24, 48, and 72 h after being sprayed with tricyclazole. In the first simulated rainfall, wash-off concentration of tricyclazole was significant irrespective of the interval between the spray time and the rainfall simulation. And from 20.5% to 24.2% of tricyclazole deposited on leaves was removed from the rice foliage. In the second simulated rainfall, concentration of tricyclazole in wash-off water was significantly lower and less than 3.6% of the deposited tricyclazole was lost.
MATERIALIEN