Merck
[Studies on chemical constituents in Huangjuhua (flowers of Chrysanthemum morifolium)].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2008-06-10)
Ya-Jun Wang, Xiu-Wwei Yang, Qiao-Sheng Guo
ZUSAMMENFASSUNG
To study the chemical constituents in Huangjuhua (flowers of Chrysanthemum morifolium). The chemical constituents were isolated by various column chromatographic methods and structurally elucidated by NMR and MS evidences. Eleven compounds were obtained and identified as luteolin (1), quercetin (2), acacetin 7-O-beta-D-(3"-acetyl) -glucopyranoside (3), luteolin 7-O-beta-D-(6"-acetyl)-glucopyranoside (4), hesperetin 7-O-beta-D-glucopyranoside (5), acacetin 7-O-beta-D-glucopyranoside (6), diosmetin 7-O-beta-D-glucopyranoside (7), apigenin 7-O-beta-D-glucopyranoside (8), hesperidin (9), linarin (10), and luteolin 7-O-beta-D-glucopyranoside (11). Compounds 4, 5 were isolated from the plant for the first time.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Apigenin-7-glucosid, analytical standard
Apigenin-7-glucosid, primary reference standard