Merck
  • [Structure-based drug development and medical/biological application of cathepsin specific inhibitors].
[Structure-based drug development and medical/biological application of cathepsin specific inhibitors].
Seikagaku. The Journal of Japanese Biochemical Society (2009-12-17)
Nobuhiko Katunuma
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
CA-074, ≥98% (HPLC)