Merck
  • Decreased susceptibility to salt-induced hypertension in subtotally nephrectomized mice lacking the vasopressin V1a receptor.
Decreased susceptibility to salt-induced hypertension in subtotally nephrectomized mice lacking the vasopressin V1a receptor.
Cardiovascular research (2010-02-02)
Ryo Oikawa, Chihiro Hosoda, Yoshihisa Nasa, Takuya Daicho, Akito Tanoue, Gozoh Tsujimoto, Norio Takagi, Kouichi Tanonaka, Satoshi Takeo
ZUSAMMENFASSUNG
By examining vasopressin V1a receptor (V1aR) knockout (KO) mice, we previously found that the V1aR is critically involved in the regulation of normal blood pressure. The present study was undertaken to elucidate the role of the V1aR in salt-induced hypertension. We compared haemodynamic responses induced by subtotal nephrectomy + salt loading in V1aR KO mice with those of wild-type (WT) controls. The time course of changes in the systolic blood pressure and heart rate during the salt loading was attenuated in the KO mice compared with that for the WT mice. The elevation of the plasma norepinephrine level caused by the subtotal nephrectomy + salt loading was also reduced in the V1aR KO mice. A V1aR antagonist markedly lowered the arterial blood pressure in the salt-loaded WT mice but not in the normotensive WT mice or in the salt-loaded or normotensive V1aR KO mice. Whereas arginine vasopressin (AVP) administered to the lateral ventricle of the brain induced pressor and tachycardiac responses accompanied by sympathetic activation in the WT mice, these events were completely abolished in the V1aR KO mice. Also, pressor and tachycardiac responses induced by intraventricularly administered hypertonic saline in the WT mice were diminished in the V1aR KO mice. Moreover, the pressor response induced by intraventricularly administered AVP was reduced in alpha(1d) adrenoceptor KO mice, whereas the tachycardiac response did not differ from that of the WT mice. These results suggest that the V1aR is involved in the elevation of arterial blood pressure caused by dietary salt and that a V1aR antagonist, in particular regarding its effect in the brain, could have significant therapeutic potential in the treatment of hypertension.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
[β-Mercapto-β,β-cyclopentamethylenepropionyl1, O-me-Tyr2, Arg8]-Vasopressin, ≥97% (HPLC)