Merck
  • Facile synthesis of zirconium phosphonate-functionalized magnetic mesoporous silica microspheres designed for highly selective enrichment of phosphopeptides.
Facile synthesis of zirconium phosphonate-functionalized magnetic mesoporous silica microspheres designed for highly selective enrichment of phosphopeptides.
Nanoscale (2011-01-26)
Jin Lu, Yan Li, Chunhui Deng
ZUSAMMENFASSUNG
In this work, we present a facile approach for the synthesis of zirconium phosphate (ZrP)-functionalized magnetic silica mesoporous microspheres for the selective enrichment of phosphopeptides. At first, magnetic mesoporous silica microspheres were prepared by directly coating mesoporous silica onto Fe3O4 magnetic microspheres, and then addition of phosphate onto the magnetic mesoporous silica microspheres was performed using 3-(trihydroxysilyl)propyl methylphosphate. The obtained phosphate-modified magnetic mesoporous microspheres were monodispersed with a mean diameter of 350 nm, and had an obvious mesoporous silica shell (∼65 nm). Finally, the resultant phosphate-functionalized magnetic mesoporous microspheres were incubated in ZrOCl2 solution with gentle stirring overnight for the loading of Zr4+ cations. The obtained Zr4+-functionalized materials were applied to the selective enrichment of phosphopeptides from both standard protein digestion and real samples. The enriched peptides were analyzed by MALDI-TOF MS and LC-ESI MS. Experimental results demonstrated that zirconium phosphonate-modified magnetic mesoporous silica microspheres show excellent potential for the selective enrichment of phosphopeptides.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zirconium(IV)-hydrogenphosphat