- Acute toxicity and withdrawal syndromes related to γ-hydroxybutyrate (GHB) and its analogues γ-butyrolactone (GBL) and 1,4-butanediol (1,4-BD).
Acute toxicity and withdrawal syndromes related to γ-hydroxybutyrate (GHB) and its analogues γ-butyrolactone (GBL) and 1,4-butanediol (1,4-BD).
Drug testing and analysis (2011-05-07)
David M Wood, Alan D Brailsford, Paul I Dargan
PMID21548140
ZUSAMMENFASSUNG
Gamma-hydroxybutyrate (GHB) has been used as a recreational drug since the 1990s and over the last few years there has been increasing use of its analogues gamma-butyrolactone (GBL) and to a lesser extent 1,4-butanediol (1,4BD). This review will summarize the literature on the pharmacology of these compounds; the patterns and management of acute toxicity associated with their use; and the clinical patterns of presentation and management of chronic dependency associated with GHB and its analogues.