Merck
  • Isolation of gentiopicroside from Gentianae Radix and its pharmacokinetics on liver ischemia/reperfusion rats.
Isolation of gentiopicroside from Gentianae Radix and its pharmacokinetics on liver ischemia/reperfusion rats.
Journal of ethnopharmacology (2011-08-23)
Wan-Ling Chang-Liao, Chao-Feng Chien, Lie-Chwen Lin, Tung-Hu Tsai
ZUSAMMENFASSUNG
Gentiopicroside (GPS) is a secoiridoid glucoside isolated from the ethanol extract of Gentianae Radix with a content of 13%, which has been used for centuries in Chinese as a digestive aid. This study investigates the pharmacokinetics of GPS and its metabolic pathway for the liver ischemia/reperfusion (I/R) in rats. The experimental animals were anesthetized intraperitoneally (i.p.) with a mixture of urethane (1.0 g/kg) and α-chloralose (0.1 g/kg). A midline laparatomy was performed and the liver hilum was gently exposed. All structures in the portal triad (hepatic artery, portal vein, and bile duct) to the left and median liver lobes were occluded with silk thread for 30 min. Ischemia was followed by a sudden reperfusion after removing the occluding threads. After 60 min reperfusion, the rats received a single intravenous 5 mg/kg dose of GPS. The area under concentration curve (AUC) was significantly increased; however, the clearance (Cl) was significantly decreased in the liver I/R rats. Furthermore, after pretreated with SKF-525A (50 mg/kg, i.p.), a cytochrome P450 (CYP) inhibitor, AUC, elimination half-life (t(1/2)) and the mean residence time (MRT) of GPS in rat blood were significantly increased, suggesting that CYP was involved in the metabolism of GPS. For the group without liver I/R, GPS was administered at doses of 5 mg/kg and 100 mg/kg intravenously and orally, respectively. The pharmacokinetic results indicated that the AUC was 565±95.1 and 1163±273 min μg/mL and the t(1/2) of GPS was 71±9 and 106±17 min after intravenous and oral administration, respectively. The oral bioavailability of GPS was 10.3±2.4% in the rats. The status of I/R might prolong the disposition of GPS, and the plasma concentration of GPS in the liver I/R injury rats was significantly increased. The increased body exposure of GPS in the treatment of liver I/R may result from the decreased metabolism of GPS mediated by CYP in the liver.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Proadifen -hydrochlorid, analytical standard, ≥95%