Merck
  • Characteristics of protection by MgADP and MgATP of α3β3γ subcomplex of thermophilic Bacillus PS3 βY341W-mutant F1-ATPase from inhibition by 7-chloro-4-nitrobenz-2-oxa-1,3-diazole support a bi-site mechanism of catalysis.
Characteristics of protection by MgADP and MgATP of α3β3γ subcomplex of thermophilic Bacillus PS3 βY341W-mutant F1-ATPase from inhibition by 7-chloro-4-nitrobenz-2-oxa-1,3-diazole support a bi-site mechanism of catalysis.
Biochemistry. Biokhimiia (2011-11-29)
Y M Milgrom
ZUSAMMENFASSUNG
MgADP and MgATP binding to catalytic sites of βY341W-α(3)β(3)γ subcomplex of F(1)-ATPase from thermophilic Bacillus PS3 has been assessed using their effect on the enzyme inhibition by 7-chloro-4-nitrobenz-2-oxa-1,3-diazole (NBD-Cl). It was assumed that NBD-Cl can inhibit only when catalytic sites are empty, and inhibition is prevented if a catalytic site is occupied with a nucleotide. In the absence of an activator, MgADP and MgATP protect βY341W-α(3)β(3)γ subcomplex from inhibition by NBD-Cl by binding to two catalytic sites with an affinity of 37 µM and 12 mM, and 46 µM and 15 mM, respectively. In the presence of an activator lauryldimethylamine-N-oxide (LDAO), MgADP protects βY341W-α(3)β(3)γ subcomplex from inhibition by NBD-Cl by binding to a catalytic site with a K(d) of 12 mM. Nucleotide binding to a catalytic site with affinity in the millimolar range has not been previously revealed in the fluorescence quenching experiments with βY341W-α(3)β(3)γ subcomplex. In the presence of activators LDAO or selenite, MgATP protects βY341W-α(3)β(3)γ subcomplex from inhibition by NBD-Cl only partially, and the enzyme remains sensitive to inhibition by NBD-Cl even at MgATP concentrations that are saturating for ATPase activity. The results support a bi-site mechanism of catalysis by F(1)-ATPases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethyldodecylamin-N-oxid -Lösung, ~30% in H2O
Sigma-Aldrich
N,N-Dimethyldodecylamin-N-oxid, BioXtra, ≥99.0% (NT)