- Synthesis of 7 alpha,12 alpha-dihydroxy-3 alpha- [2-(beta-D-glucopyranosyl)acetyl]-5 beta-cholan-24-oic acid.
Synthesis of 7 alpha,12 alpha-dihydroxy-3 alpha- [2-(beta-D-glucopyranosyl)acetyl]-5 beta-cholan-24-oic acid.
Steroids (1990-07-01)
P Allevi, M Anastasia, P Ciuffreda, A Fiecchi, A M Sanvito
PMID2219197
ZUSAMMENFASSUNG
A C-glucoside of cholic acid was synthesized by the introduction of an acetyl group at position 3 alpha and direct one-pot C-glucosidation using 2,3,4,6-tetra-O-benzyl-alpha-D-glucopyranosyl chloride.