Merck
Levosimendan protection against kidney ischemia/reperfusion injuries in anesthetized pigs.
The Journal of pharmacology and experimental therapeutics (2012-05-09)
Elena Grossini, Claudio Molinari, Piero Pollesello, Giorgio Bellomo, Guido Valente, David Mary, Giovanni Vacca, Philippe Caimmi
ZUSAMMENFASSUNG
Ischemia/reperfusion (I/R) injury is an important cause of acute renal failure because of oxidative, inflammatory, and apoptotic mechanisms. The aim of the present study was to examine any possible protective effects of levosimendan in an in vivo pig model of renal I/R injury. In 40 anesthetized pigs (eight groups of five pigs each), I/R was induced by clamping-reopening the left renal artery. During ischemia, in three groups of pigs, levosimendan and the multiorgan preservation solution Custodiol, alone or in combination with levosimendan, were infused in the renal artery. In two other groups of animals, levosimendan in combination with Custodiol was administered after the intrarenal nitric-oxide (NO) synthase blocker N(ω)-nitro-L-arginine methyl ester (L-NAME) or the mitochondrial ATP-sensitive K(+) channel (K(ATP) channel) inhibitor 5-hydroxydecanoate (5-HD). In the other animals, saline, L-NAME, or 5-HD were administered alone. Throughout the experiments, urinary N-acetyl-β-glucosaminidase (NAG) release was measured, and renal function was assessed. Moreover, renal biopsy samples were taken for the detection of apoptosis and tissue peroxidation. In pigs treated with levosimendan or the combination of levosimendan and Custodiol, NAG, peroxidation, and apoptotic markers were lower than in animals treated with Custodiol alone. In addition, renal function was better preserved, and cell survival and antioxidant systems were more activated. All beneficial effects were prevented by L-NAME and 5-HD. In conclusion, levosimendan alone or in combination with Custodiol exerted better protection against renal I/R injuries than Custodiol alone through antioxidant, antiapoptotic, and prosurvival actions depending on mitochondrial K(ATP) channels and NO-related mechanisms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5-Hydroxydecanoic acid sodium salt, ≥97% (HPLC)