Merck
  • Characterization of a newly isolated highly effective 3,5,6-trichloro-2-pyridinol degrading strain Cupriavidus pauculus P2.
Characterization of a newly isolated highly effective 3,5,6-trichloro-2-pyridinol degrading strain Cupriavidus pauculus P2.
Current microbiology (2012-05-31)
Li Cao, Hongming Liu, Hao Zhang, Ke Huang, Tao Gu, Haiyan Ni, Qing Hong, Shunpeng Li
ZUSAMMENFASSUNG
A bacterial strain P2 capable of degrading 3,5,6-trichloro-2-pyridinol (TCP) was isolated and characterized. Phylogenetic analysis based on 16S rRNA gene sequence indicated that it belonged to the genus of Cupriavidus, because it showed the highest sequence similarity to Cupriavidus pauculus LMG 3413(T) (99.7 %) and DNA-DNA relatedness value between strain P2 and C. pauculus LMG 3413(T) was 76.8 %. In combination with morphological, physiological and biochemical characters, strain P2 was identified as C. pauculus. It could use TCP as the sole carbon source and energy source for its growth. It showed a high average degradation rate of 10 mg/L h in mineral salt medium amended with TCP (50-800 mg/L). During TCP degradation, chloridion was released into the medium in two obvious discontinuous stages. Along with this, two colorful metabolites were produced. Finally, the molarity of the total released chloridion was three times that of the initial TCP in the medium. This is the first report of TCP-degrading strain from the genus of Cupriavidus and the detection of two colorful metabolites during TCP degradation. Strain P2 might be a promising candidate for its application in the bioremediation of TCP-polluted environments.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
3,5,6-Trichlor-2-pyridinol, PESTANAL®, analytical standard