Merck
  • C-Glucosylated malonitrile as a key intermediate towards carbohydrate-based glycogen phosphorylase inhibitors.
C-Glucosylated malonitrile as a key intermediate towards carbohydrate-based glycogen phosphorylase inhibitors.
Bioorganic & medicinal chemistry (2012-08-23)
Sophie Feuillastre, Aikaterini S Chajistamatiou, Constantinos Potamitis, Maria Zervou, Panagiotis Zoumpoulakis, Evangelia D Chrysina, Jean-Pierre Praly, Sébastien Vidal
ZUSAMMENFASSUNG
Glycogen utilization involves glycogen phosphorylase, an enzyme which appears to be a potential target for the regulation of glycaemia, as the liver isoform is a major player for hepatic glucose output. A single C-glucosylated malonitrile allowed for the synthesis of three glucose-based derivatives namely bis-oxadiazoles, bis-amides and a C-glucosylated tetrahydropyrimidin-2-one. When evaluated as glycogen phosphorylase inhibitors, two of the synthesized compounds displayed inhibition in the sub-millimolar range. In silico studies revealed that only one out of the bis-amides obtained and the C-glucosylated tetrahydropyrimidin-2-one may bind at the catalytic site.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Malonsäuredinitril, ≥99%
Sigma-Aldrich
Malonsäuredinitril, Arxada quality, ≥99.0% (calculated, GC, KF)