Merck
Dioscin: a synergistic tyrosinase inhibitor from the roots of Smilax china.
Food chemistry (2012-10-31)
Chun Liang, Ju-Hwan Lim, Seung-Hyung Kim, Dong-Seon Kim
ZUSAMMENFASSUNG
In our study, we investigated the inhibition of mushroom tyrosinase in Smilax china. A methanol (MeOH) extract of S. china was partitioned into hexane, ethyl acetate (EtOAc) and water. Of the three fractions, EtOAc extract showed the strongest inhibition of tyrosinase activity with l-tyrosine or l-DOPA as a substrate. Two compounds were isolated from a final active fraction by activity-guided column chromatography. These compounds were identified as dioscin and oxyresveratrol by comparing their mass, (1)H- and (13)C-NMR spectral data with those reported in the literature. Dioscin showed little inhibition activity of tyrosinase, whereas oxyresveratrol, a known tyrosinase inhibitor, showed a strong tyrosinase inhibitory activity. We discovered that a mixture of oxyresveratrol and dioscin (IC(50)=5.1 and 5.7 μg/ml) highly increased the inhibition of tyrosinase activity with l-tyrosine or l-DOPA as the substrate as compared to either oxyresveratrol (IC(50)=7.8 and 10.9 μg/ml) or dioscin (IC(50)>100 and 100 μg/ml) alone.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Diosgenin, ≥93%