Merck
  • Expression of Paracoccidioides brasiliensis AMY1 in a Histoplasma capsulatum amy1 mutant, relates an α-(1,4)-amylase to cell wall α-(1,3)-glucan synthesis.
Expression of Paracoccidioides brasiliensis AMY1 in a Histoplasma capsulatum amy1 mutant, relates an α-(1,4)-amylase to cell wall α-(1,3)-glucan synthesis.
PloS one (2012-11-28)
Emma Camacho, Victoria E Sepulveda, William E Goldman, Gioconda San-Blas, Gustavo A Niño-Vega
ZUSAMMENFASSUNG
In the cell walls of the pathogenic yeast phases of Paracoccidioides brasiliensis, Blastomyces dermatitidis and Histoplasma capsulatum, the outer α-(1,3)-glucan layer behaves as a virulence factor. In H. capsulatum, an α-(1,4)-amylase gene (AMY1) is essential for the synthesis of this polysaccharide, hence related to virulence. An orthologous gene to H. capsulatum AMY1 was identified in P. brasiliensis and also labeled AMY1. P. brasiliensis AMY1 transcriptional levels were increased during the yeast phase, which correlates with the presence of α-(1,3)-glucan as the major yeast cell wall polysaccharide. Complementation of a H. capsulatum amy1 mutant strain with P. brasiliensis AMY1, suggests that P. brasiliensis Amy1p may play a role in the synthesis of cell wall α-(1,3)-glucan. To study some biochemical properties of P. brasiliensis Amy1p, the enzyme was overexpressed, purified and studied its activity profile with starch and amylopeptin. It showed a relatively higher hydrolyzing activity on amylopeptin than starch, producing oligosaccharides from 4 to 5 glucose residues. Our findings show that P. brasiliensis Amy1p produces maltooligosaccharides which may act as a primer molecule for the fungal cell wall α-(1,3)-glucan biosynthesis by Ags1p.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use in purification of Histidine-tagged proteins, DMSO solution
Sigma-Aldrich
Menadione sodium bisulfite, ≥95% (TLC)
Sigma-Aldrich
Amylopectin aus Mais
Sigma-Aldrich
Amylopektin aus Kartoffelstärke