Merck
Microchip UV absorbance detection applied to isoelectric focusing of proteins.
Methods in molecular biology (Clifton, N.J.) (2013-01-19)
Junjie Ou, Carolyn L Ren
ZUSAMMENFASSUNG
Isoelectric focusing (IEF) is considered as an attractive separation technique for biologically amphoteric compounds (e.g., proteins and peptides) based on their isoelectric point (pI). With the advancement in micromachining technology, microchip format IEF has attracted significant attention. Both single-point and whole column imaging detection (WCID) methods have been employed for analyzing the separation performance in a microchip. WCID is more favorable than single-point detection because the latter requires the focused bands to be mobilized and thus adds more complexity to the design and operation of such microchips. Fluorescence- and UV absorbance-based WCID have been successfully adapted in glass and PDMS microchips. We have developed polydimethylsiloxane (PDMS) microchips for IEF applications where UV-WCID is employed for evaluating the separation performance. The chips are designed for use in the iCE280 analyzer (Convergent Bioscience Inc., Toronto), for capillary-based IEF where UV-WCID is employed for analyzing the separation performance. Three kinds of microchips that have been successfully developed using standard soft lithography technology are described in detail.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Siliconöl, viscosity 5 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 10 cSt (25 °C)
Sigma-Aldrich
Siliconöl
Sigma-Aldrich
Siliconöl, viscosity 100 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 20 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 1,000 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 50 cSt (25 °C)
Siliconöl, for melting point and boiling point apparatuses
Sigma-Aldrich
Siliconöl, viscosity 500 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 350 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 10,000 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 60,000 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 30,000 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 100,000 cSt (25 °C)