Merck
  • Synergistic antibacterial activity in vitro of trimethoprim and sulphonamides and the importance of pharmacokinetic properties for optimal therapy.
Synergistic antibacterial activity in vitro of trimethoprim and sulphonamides and the importance of pharmacokinetic properties for optimal therapy.
Infection (1979-01-01)
J K Seydel, E Wempe
PMID389807