Merck
  • Cyclosporine A and Rapamycin induce in vitro cholesteryl ester transfer protein activity, and suppress lipoprotein lipase activity in human plasma.
Cyclosporine A and Rapamycin induce in vitro cholesteryl ester transfer protein activity, and suppress lipoprotein lipase activity in human plasma.
International journal of pharmaceutics (2008-05-02)
Rita Tory, Kristina Sachs-Barrable, John S Hill, Kishor M Wasan
ZUSAMMENFASSUNG
Cyclosporine A (CsA), Rapamycin (RAPA), Tacrolimus (FK-506) and Mycophenolate mofetil (MMF) are immunosuppressants that are widely used in solid organ transplant patients. However, some of these drugs have been reported to cause dyslipidemia in patients. Our aim was to determine the effects of these drugs on in vitro cholesteryl ester transfer protein (CETP), hepatic lipase (HL) and lipoprotein lipase (LPL) activity within human plasma. We measured CETP activity in human normolipidemic plasma with and without drug treatment, by measuring the incorporation of labeled cholesteryl ester into lipoproteins. To further confirm the result, we also measured recombinant CETP (rCETP) activity with and without drug treatment. We measured HL and LPL activity in post-heparin normal human plasma in the presence and absence of the drugs by measuring the release of fatty acids from radiolabeled triolein. We found an increase in CETP activity in human normolipidemic plasma and rCETP treated with CsA and RAPA. By contrast, CETP activity was not altered significantly in the presence of FK-506 and MMF. LPL activity in post-heparin normal human plasma was suppressed following the co-incubation with CsA, RAPA, FK-506 or MMF whereas HL activity remained unaffected. The increase in CETP activity and suppression in LPL activity following CsA and RAPA treatment observed in the present study may be associated with elevated LDL cholesterol levels and hypertriglyceridemia seen in patients administered these drugs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
CETP Activity Assay Kit, Supplied by Roar Biomedical, Inc.