Merck
  • Triton X-100 concentration effects on membrane permeability of a single HeLa cell by scanning electrochemical microscopy (SECM).
Triton X-100 concentration effects on membrane permeability of a single HeLa cell by scanning electrochemical microscopy (SECM).
Proceedings of the National Academy of Sciences of the United States of America (2010-09-15)
Dipankar Koley, Allen J Bard
ZUSAMMENFASSUNG
Changes in HeLa cell morphology, membrane permeability, and viability caused by the presence of Triton X-100 (TX100), a nonionic surfactant, were studied by scanning electrochemical microscopy (SECM). No change in membrane permeability was found at concentrations of 0.15 mM or lower during an experimental period of 30 to 60 min. Permeability of the cell membrane to the otherwise impermeable, highly charged hydrophilic molecule ferrocyanide was seen starting at concentrations of TX100 of about 0.17 mM. This concentration level of TX100 did not affect cell viability. Based on a simulation model, the membrane permeability for ferrocyanide molecules passing though the live cell membrane was 6.5 ± 2.0 × 10(-6) m/s. Cells underwent irreversible permeabilization of the membrane and structural collapse when the TX100 concentration reached the critical micelle concentration (CMC), in the range of 0.19 to 0.20 mM. The impermeability of ferrocyanide molecules in the absence of surfactant was also used to determine the height and diameter of a single living cell with the aid of the approach curve and probe scan methods in SECM.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade