- Enantioselective binding sites on bovine serum albumin to dansyl amino acids.
Enantioselective binding sites on bovine serum albumin to dansyl amino acids.
Biochimica et biophysica acta (1999-08-14)
Y Abe, S Fukui, Y Koshiji, M Kobayashi, T Shoji, S Sugata, H Nishizawa, H Suzuki, K Iwata
PMID10446371
ZUSAMMENFASSUNG
The enantioselective binding sites on bovine serum albumin were examined by HPLC using 19 racemic 5-N, N-dimethylamino-1-naphthalenesulfonyl derivatives of alpha-amino acids (dansyl amino acids) as chiral probes. On a bovine serum albumin bonded chiral stationary phase, seven L-forms eluted faster than their D-forms, while ten D-forms eluted before their L-forms. It was speculated that either two classes or two different binding sites exist on bovine serum albumin which can be distinguished by N-dansyl-L-proline and N-dansyl-D-norvaline. This was confirmed by fluorometric experiments where non-fluorescent 1-naphthalenesulfonyl derivatives were synthesized and competitive adsorption experiments were performed.