Merck
Bismuth subgallate-epinephrine paste in adenotonsillectomies.
The Annals of pharmacotherapy (2000-04-20)
R C Hatton
ZUSAMMENFASSUNG
To evaluate the role of bismuth subgallate-epinephrine (BSE) paste as a hemostatic in adenotonsillectomies. MEDLINE (January 1966-October 1999) and Current Contents (January 1997-October 1999) were searched, using bismuth subgallate, adenoidectomy, tonsillectomy, and adenotonsillectomy as search terms. A citation search was performed using Science Citation Index (January 1977-October 1999). Adenotonsillectomies are common procedures; although there are few complications, hemorrhage is a concern. Bismuth subgallate has historically been used as an astringent and hemostatic. An evaluation of studies of bismuth subgallate and BSE paste was conducted. There is minimal evidence to support this practice, but data suggest that epinephrine may be the active ingredient in BSE paste. BSE paste is inexpensive, poses little risk, and may decrease postoperative bleeding; therefore, it may be a reasonable hemostatic agent.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bismut(III)-gallat basisch Hydrat, ≥98.0% (calc. on dry substance, KT)