Merck
Limitations of dextromethorphan N-demethylation as a measure of CYP3A activity.
Pharmacogenetics (2000-04-26)
A D Kashuba, A N Nafziger, G L Kearns, J S Leeder, R Gotschall, A Gaedigk, J S Bertino
ZUSAMMENFASSUNG
We evaluated the utility of the 3-methoxymorphinan/dextromethorphan (3MM/DM) urinary ratio to reflect baseline CYP3A activity, and its ability to discriminate moderate CYP3A inhibition during fluvoxamine therapy. For 4 months, oral dextromethorphan 30 mg and intravenous midazolam 0.025 mg/kg were administered to nine men every 14 days, and to 10 premenopausal women during the follicular and luteal phases of their menstrual cycles. Phenotyping during the first 3 months or cycles established baseline CYP3A activity. During the fourth month, individuals were given fluvoxamine 150 mg/day. CYP3A activity was expressed as both the urinary 3MM/DM molar ratio and midazolam plasma clearance (MDZ CL). 3MM/DM ratios were independent of dextromethorphan CYP2D6 phenotype (r = 0.13, P = 0.6). Intraindividual variability in baseline CYP3A activity (median, 25-75th percentile), as determined by coefficients of variation, was 48.3% (36.8-68.8%) for 3MM/DM and 10.3% (8.3-11.8%) for MDZ CL. No significant correlation between 3MM/DM and MDZ CL either at baseline (r = -0.22, P = 0.4) or during fluvoxamine therapy (r = -0.15, P = 0.6) was noted. With fluvoxamine 150 mg/day, median percentage change in the 3MM/DM ratios was -50.0% (-105.6-6.0%; P = 0.7), and median percentage change in MDZ CL was -33.7% (-27.0-39.3%; P < 0.0001). Only MDZ CL consistently indicated moderate inhibition of hepatic CYP3A activity. In addition, there was a lack of correlation between the magnitudes of fluvoxamine-induced change in 3MM/DM and MDZ CL (r = 0.41, P = 0.1). The large intraindividual variability of the 3MM/DM urinary ratio, as well as the inability to discriminate moderate CYP3A inhibition, makes this a suboptimal method for accurately assessing CYP3A activity.