Merck
Pelorol from the tropical marine sponge Dactylospongia elegans.
Journal of natural products (2000-09-09)
E Goclik, G M König, A D Wright, R Kaminsky
ZUSAMMENFASSUNG
From the dichloromethane solubles of the tropical marine sponge Dactylospongia elegans, a new aromatic substituted sesquiterpene, pelorol (1), and the known sesquiterpene, ilimaquinone (2), were isolated. The structures of 1 and 2 were deduced from their spectroscopic data. The biological activities of compounds 1 and 2 were assessed in a variety of bioassays, and both compounds were found to have weak antitrypanosomal and antiplasmodial effects.