Merck
  • Separation of the four pairs of enantiomers of vincamine alkaloids by enantioselective high-performance liquid chromatography.
Separation of the four pairs of enantiomers of vincamine alkaloids by enantioselective high-performance liquid chromatography.
Journal of chromatography. A (2000-10-24)
S Caccamese, G Principato
ZUSAMMENFASSUNG
The four enantiomeric pairs of vincamine group alkaloids were separated by HPLC using Chiralpak AD as chiral stationary phase (CSP) and various n-hexane-2-propanol and n-hexane-ethanol mobile phases. (+)-cis-Vincamine, which is used in pharmaceutical preparations, is eluted much faster than its optical isomer, with separation factors of 2.4 and 3.5, respectively in these mobile phases. Other CSPs gave negative results. A chiral recognition mechanism is proposed and circular dichroism spectra of the individual enantiomers are presented.