Merck
  • Simultaneous up-regulation of urokinase-type plasminogen activator (uPA) and uPA receptor by hepatocyte growth factor/scatter factor in human glioma cells.
Simultaneous up-regulation of urokinase-type plasminogen activator (uPA) and uPA receptor by hepatocyte growth factor/scatter factor in human glioma cells.
Clinical & experimental metastasis (2000-11-23)
T Moriyama, H Kataoka, R Hamasuna, E Yoshida, T Sameshima, T Iseda, K Yokogami, S Nakano, M Koono, S Wakisaka
ZUSAMMENFASSUNG
Several lines of evidence indicate that hepatocyte growth factor/scatter factor (HGF/SF) and its receptor, c-Met, may play an important role in progression of human glioma. In this study, effects of HGF/SF on urokinase- type plasminogen activator (uPA)-mediated proteolysis network were examined in c-Met-positive human glioma cell lines. Treatment of the glioma cells with various concentrations of HGF/SF resulted in an enhanced secretion of uPA proteins accompanying increased transcription of uPA mRNA in a dose dependent fashion. The levels of uPA receptor (uPAR) mRNAs were also elevated simultaneously upon HGF/SF stimulation, and the cell-surface associated uPA activity was also elevated by the treatment. Since concomitant expression of HGF and its receptor c-Met are frequently observed in malignant gliomas, these results suggest that HGF/SF participates in invasive process of malignant glioma cells not only by its motility-stimulating activity but also through enhanced degradation of the extracellular matrix induced by autocrine activation of uPA proteolysis network.