Merck
  • Application of ferrous hydrogen peroxide for treatment of DSD-acid manufacturing process wastewater.
Application of ferrous hydrogen peroxide for treatment of DSD-acid manufacturing process wastewater.
Water research (2001-05-08)
W Zhu, Z Yang, L Wang
ZUSAMMENFASSUNG
A pretreatment method for the biological treatment of wastewater from 4,4'-diaminostilbene-2,2'-disulfonic acid (DSD-acid) manufacturing processes, a refractory dye intermediate wastewater, based on combined ferrous hydrogen peroxide oxidation and coagulation-flocculation, was developed. When the wastewater was treated with ferrous hydrogen peroxide oxidation ([Fe2+] = 2.7 mmol/L, [H2O2] = 0.21 mol/L) after a flocculation using an organic flocculant TS-1 at a dosage of 3 g/L, the overall COD and color removals were 64 and 62%, respectively. BOD5/COD value of the effluent was 0.3. Ferrous hydrogen peroxide oxidation treatment can reduce the solubility of organic molecules with sulfonic group and increase the efficiency of coagulation treatment. The COD and color removals were both more than 90% when FeCl3 was used as the coagulation (dosages of two-step coagulation were 0.031 and 0.012 mol/L respectively) after a ferrous hydrogen peroxide oxidation pretreatment at a H2O2 dosage of 0.06 mol/L.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4,4′-Diamino-2,2′-stilbendisulfonsäure, technical grade, 85%