Merck
  • Combined treatment with NMDA antagonist, CGP 37849, and sigma receptor agonists, SA4503 or DTG, decreases the neuroleptic-induced catalepsy in rats.
Combined treatment with NMDA antagonist, CGP 37849, and sigma receptor agonists, SA4503 or DTG, decreases the neuroleptic-induced catalepsy in rats.
Polish journal of pharmacology (2001-05-10)
G Skuza
ZUSAMMENFASSUNG
The obtained results indicate that joint administration of CGP 37849, a competitive NMDA receptor antagonist, and SA4503, a sigma1 (sigma1) receptor agonist or DTG, sigma 1/2 receptor agonist, evoked anticataleptic effect at doses which were ineffective when each of the compounds was given alone.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
CGP 37849, ≥98% (HPLC)