Merck
  • Mutagenicity of bay-region amino-substituted cyclopenta[a]phenanthrenes and 2- and 5-aminochrysene.
Mutagenicity of bay-region amino-substituted cyclopenta[a]phenanthrenes and 2- and 5-aminochrysene.
Mutation research (2001-05-30)
F S Catterall, M M Coombs, C Ioannides, J J Sepiol, J Wilson
ZUSAMMENFASSUNG
The relative mutagenic potentials of 11-amino-16,17-dihydro-15H-cyclopenta[a]phenanthrene, its 17-keto derivative, and 2- and 5-aminochrysene have been compared in Salmonella typhimurium TA98 and TA100 in the presence of a postmitochondrial liver preparation from Aroclor 1254 induced rats. The 11-amino hydrocarbon is a very weak mutagen (0.27 revertants/nmol), whereas the 11-amino-17-ketone is much more active (129 revertants/nmol). 2-Aminochrysene is the most mutagenic arylamine ( approximately 500 revertants/nmol) among these compounds, but its 5-amino isomer is much less active (0.9 revertants/nmol). Possible reasons for these marked differences are suggested. Use of TA98 with over-expressing O-acetyltransferase (YG 1024) and deficient in this enzyme (TA98/l,8-DNP(6)) with the 11-amino-17-ketone and with 5-aminochrysene clearly indicates the importance of this enzyme in their bioactivation, implying oxidation of the amino group to the hydroxylamine in both these compounds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
6-Aminochrysen, 95%